2-Bromopyridine ≥99%
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 109-04-6 Molecular Formula: C5H4BrN Formula Weight: 158.00 Flash Point: 97°(207°F) Refractive Index: 1.5720 Odour: Characteristic Boiling Point: 192-194° Beilstein Reference: 105789 UN No.: 2810 MDL No.: MFCD00006219
- CAS Number:109-04-6
- Formula:C₅H₄BrN
- MDL Number:MFCD00006219
- Molecular Weight:158 g/mol
- Boiling Pt:192…194 °C
- Melting Pt:–40.1 °C
- Density:1.625
- Flash Pt:97 °C (207 °F)
- Storage Temperature:Ambient
- EINECS:203-641-6
- UN:2810
- ADR:6.1,II
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



