Order Entry
Export
Orders LinkContactUsLinkComponent
Ethyl 4-hydroxybenzoate ≥99%

Ethyl 4-hydroxybenzoate ≥99%

Glass bottle for solids, 5.000 g
Catalog # A13172.0I
Ethyl 4-hydroxybenzoate ≥99%
Catalog # A13172.0I
CAS Number:  120-47-8
Ethyl 4-hydroxybenzoate  ≥99%
Ethyl 4-hydroxybenzoate ≥99%

Glass bottle for solids, 5.000 g

CAS Number:  120-47-8
Price
412,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents

Purity: 99% CAS No.: 120-47-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 116-118° Boiling Point: 297-298° dec. Merck Reference: 14,3837 MDL No.: MFCD00002353

Synonyms:  Nipagin A, Ethyl paraben, Ethyl p-hydroxybenzoate, Ethylparaben, Ethyl 4-hydroxybenzoate
  • CAS Number:120-47-8
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00002353
  • Molecular Weight:166.18 g/mol
  • Merck Index:14,3837
  • Boiling Pt:297…298 °C (dec.)
  • Melting Pt:116…118 °C
  • Density:1.078 g/cm³ (131 °C)
  • Storage Temperature:Ambient
  • EINECS:204-399-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications