Order Entry
Export
Orders LinkContactUsLinkComponent
2-Fluoro-4-nitrophenol ≥98%

2-Fluoro-4-nitrophenol ≥98%

Glass bottle for solids, 25 g
  A13134.14
Toxic
Irritant
2-Fluoro-4-nitrophenol ≥98%
  A13134.14
 :  403-19-0
2-Fluoro-4-nitrophenol ≥98%
2-Fluoro-4-nitrophenol ≥98%

Glass bottle for solids, 25 g

 :  403-19-0
 
256,00 €
Log in for availability
1 * 25 g
 

Purity: 98+% CAS No.: 403-19-0 Molecular Formula: C6H4FNO3 Formula Weight: 157.10 Melting Point: 117-121° Beilstein Reference: 1944995

  • CAS Number:403-19-0
  • Formula:C₆H₄FNO₃
  • MDL Number:MFCD00051970
  • Molecular Weight:157.1 g/mol
  • Melting Pt:117…121 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications