Order Entry
Export
Orders LinkContactUsLinkComponent
Methyl methacrylate 99% stabilised

Methyl methacrylate 99% stabilised

Glass bottle, 100 mL
Catalog # A13030.AE
Flammable
Irritant
Methyl methacrylate 99% stabilised
Catalog # A13030.AE
CAS Number:  80-62-6
Methyl methacrylate 99% stabilised
Methyl methacrylate 99% stabilised

Glass bottle, 100 mL

CAS Number:  80-62-6
Price
26,70 €
Log in for availability
1 * 100 mL
Product Details & Documents

Stabilized

Purity: 99% CAS No.: 80-62-6 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 10°(50°F) Refractive Index: 1.4150 Melting Point: -48° Boiling Point: 100-101° Beilstein Reference: 605459 Merck Reference: 14,5941 UN No.: 1247 MDL No.: MFCD00008587

Synonyms:  Methacrylic acid methyl ester
  • CAS Number:80-62-6
  • Formula:CH₂=C(CH₃)COOCH₃
  • MDL Number:MFCD00008587
  • Molecular Weight:100.12 g/mol
  • Merck Index:14,5941
  • Boiling Pt:100 °C (760 mmHg)
  • Melting Pt:–48 °C
  • Density:0.93 g/cm³
  • Flash Pt:8 °C
  • UN:1247
  • ADR:3,II
  •  :Stabilised with hydroquinone (HQ)
  • Storage Temperature:Ambient
  • EINECS:201-297-1
  • REACH:01-2119452498-28
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications