Order Entry
Export
Orders LinkContactUsLinkComponent
4-Iodopyridine 96%

4-Iodopyridine 96%

Glass bottle for solids, 1 g
Catalog # B25459.03
Irritant
4-Iodopyridine 96%
Catalog # B25459.03
CAS Number:  15854-87-2
4-Iodopyridine 96%
4-Iodopyridine 96%

Glass bottle for solids, 1 g

CAS Number:  15854-87-2
Price
53,40 €
Log in for availability
1 * 1 g
Product Details & Documents

Purity: 96% CAS No.: 15854-87-2 Molecular Formula: C5H4IN Formula Weight: 205.00 Storage Temperature: 2°C to 8°C Melting Point: 96-99° Beilstein Reference: 1594 MDL No.: MFCD02093937

  • CAS Number:15854-87-2
  • Formula:C₅H₄IN
  • MDL Number:MFCD02093937
  • Molecular Weight:205 g/mol
  • Melting Pt:96…99 °C
  • Storage Temperature:Freezer
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications