Order Entry
Export
Orders LinkContactUsLinkComponent
Nalidixic acid ≥99%

Nalidixic acid ≥99%

Glass bottle for solids, 100 g
Catalog # B25096.22
Irritant
Nalidixic acid ≥99%
Catalog # B25096.22
CAS Number:  389-08-2
Nalidixic acid ≥99%
Nalidixic acid ≥99%

Glass bottle for solids, 100 g

CAS Number:  389-08-2
Price
390,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 389-08-2 Molecular Formula: C12H12N2O3 Formula Weight: 232.24 Melting Point: 228-230° Beilstein Reference: 750515 Merck Reference: 14,6359 MDL No.: MFCD00006884

  • CAS Number:389-08-2
  • Formula:C₁₂H₁₂N₂O₃
  • MDL Number:MFCD00006884
  • Molecular Weight:232.24 g/mol
  • Merck Index:14,6359
  • Melting Pt:228…230 °C
  • Storage Temperature:Ambient
  • EINECS:206-864-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications