Order Entry
Export
Orders LinkContactUsLinkComponent
1,1-Dimethoxyacetone ≥97%

1,1-Dimethoxyacetone ≥97%

Glass bottle for solids, 500 g
Catalog # B24988.36
Flammable
Irritant
1,1-Dimethoxyacetone ≥97%
Catalog # B24988.36
CAS Number:  6342-56-9
1,1-Dimethoxyacetone ≥97%
1,1-Dimethoxyacetone ≥97%

Glass bottle for solids, 500 g

CAS Number:  6342-56-9
Price
143,00 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 97+% CAS No.: 6342-56-9 Molecular Formula: CH3COCH(OCH3)2 Formula Weight: 118.13 Flash Point: 34°(93°F) Refractive Index: 1.3980 Melting Point: -57° Boiling Point: 133-135° Beilstein Reference: 1560557 UN No.: 1993 MDL No.: MFCD00008758

Synonyms:  Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
  • CAS Number:6342-56-9
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00008758
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:133…135 °C
  • Melting Pt:–57 °C
  • Density:0.998
  • Flash Pt:34 °C (93 °F)
  • Storage Temperature:Ambient
  • EINECS:228-735-4
  • UN:1993
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications