Methyl-2,6-dichlorobenzoate 97%
25 g
Catalog # B24912.14
Supplier: Thermo Fisher Scientific
Methyl-2,6-dichlorobenzoate 97%
Catalog # B24912.14
Supplier: Thermo Fisher Scientific
CAS Number: 14920-87-7
Jump to:
Product Details & Documents
Synonyms: Methyl 2,6-dichlorobenzoate
- CAS Number:14920-87-7
- Formula:C₈H₆Cl₂O₂
- MDL Number:MFCD01313701
- Molecular Weight:205.04
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pk:25 g



