Order Entry
Export
Orders LinkContactUsLinkComponent
 

Methyl-2,6-dichlorobenzoate 97%

1 g
Catalog # B24912.03
Methyl-2,6-dichlorobenzoate 97%
Catalog # B24912.03
CAS Number:  14920-87-7
Methyl-2,6-dichlorobenzoate 97%
Methyl-2,6-dichlorobenzoate 97%

1 g

CAS Number:  14920-87-7
Price
28,40 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Methyl 2,6-dichlorobenzoate
  • CAS Number:14920-87-7
  • Formula:C₈H₆Cl₂O₂
  • MDL Number:MFCD01313701
  • Molecular Weight:205.04
Specifications

Specifications

  • Pk:
    1 g

Specifications