Order Entry
Export
Orders LinkContactUsLinkComponent
1-Tridecene ≥97%

1-Tridecene ≥97%

Glass bottle for solids, 5 g
Catalog # B24413.06
Health
1-Tridecene ≥97%
Catalog # B24413.06
CAS Number:  2437-56-1
1-Tridecene ≥97%
1-Tridecene ≥97%

Glass bottle for solids, 5 g

CAS Number:  2437-56-1
Price
71,10 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 2437-56-1 Molecular Formula: C13H26 Formula Weight: 182.35 Flash Point: 79°(174°F) Refractive Index: 1.4330 Odour: Mild Melting Point: -23° Boiling Point: 232-233° Beilstein Reference: 1744660 MDL No.: MFCD00008976

  • CAS Number:2437-56-1
  • Formula:C₁₃H₂₆
  • MDL Number:MFCD00008976
  • Molecular Weight:182.35 g/mol
  • Boiling Pt:232…233 °C
  • Melting Pt:–23 °C
  • Density:0.766
  • Flash Pt:79 °C (174 °F)
  • Storage Temperature:Ambient
  • EINECS:219-443-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications