1,5-Diaminonaphthalene 97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
1,5-Diaminonaphthalene 97%
Catalog # B24262.06
Supplier: Thermo Fisher Scientific
CAS Number: 2243-62-1
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 2243-62-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 189-192° Boiling Point: 200-210°/5mm UN No.: 3077 MDL No.: MFCD00004029
Synonyms: 1,5-naphthylenediamine, 1,5-Naphthalenediamine
- CAS Number:2243-62-1
- Formula:C₁₀H₁₀N₂
- MDL Number:MFCD00004029
- Molecular Weight:158.2 g/mol
- Boiling Pt:200…210 °C (5 mmHg)
- Melting Pt:189…192 °C
- Density:1.4 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:218-817-8
- UN:3077
- ADR:9,III
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



