Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,3-Dibromobutane ≥98%

Glass bottle for solids, 100 g
Catalog # B24123.22
Irritant
1,3-Dibromobutane ≥98%
Catalog # B24123.22
CAS Number:  107-80-2
1,3-Dibromobutane ≥98%
1,3-Dibromobutane ≥98%

Glass bottle for solids, 100 g

CAS Number:  107-80-2
Price
73,20 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98% CAS No.: 107-80-2 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: None Refractive Index: 1.5095 Boiling Point: 173-176° Beilstein Reference: 1731231 MDL No.: MFCD00000152

  • CAS Number:107-80-2
  • Formula:BrH₂CCH₂BrCHCH₃
  • MDL Number:MFCD00000152
  • Molecular Weight:215.92 g/mol
  • Boiling Pt:173…176 °C
  • Density:1.789
  • Storage Temperature:Ambient
  • EINECS:203-520-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications