Orders LinkContactUsLinkComponent
1,3-Dibromobutane ≥98%
1,3-Dibromobutane ≥98%
Glass bottle for solids, 25 g
  B24123.14
1,3-Dibromobutane ≥98%
  B24123.14
 :  107-80-2
1,3-Dibromobutane ≥98%
1,3-Dibromobutane ≥98%
Glass bottle for solids, 25 g
 :  107-80-2
28,80 €
1 * 25 g
Purity: 98% CAS No.: 107-80-2 Molecular Formula: C4H8Br2 Formula Weight: 215.93 Flash Point: None Refractive Index: 1.5095 Boiling Point: 173-176° Beilstein Reference: 1731231 MDL No.: MFCD00000152
  •  :107-80-2
  •  :BrH₂CCH₂BrCHCH₃
  •  :MFCD00000152
  •  :215.92 g/mol
  •  :173…176 °C
  •  :1.789
  •  :Ambient
  •  :203-520-8
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g