Order Entry
Export
Orders LinkContactUsLinkComponent
Catechol violet

Catechol violet

Glass bottle for solids, 5 g
Catalog # B23513.06
Irritant
Catechol violet
Catalog # B23513.06
CAS Number:  115-41-3
Catechol violet
Catechol violet

Glass bottle for solids, 5 g

CAS Number:  115-41-3
Price
54,10 €
Log in for availability
1 * 5 g
Product Details & Documents

CAS No.: 115-41-3 Molecular Formula: C19H14O7S Formula Weight: 386.38 Beilstein Reference: 353938 MDL No.: MFCD00005868

Synonyms:  Pyrocatechol Violet
  • CAS Number:115-41-3
  • Formula:C₁₉H₁₄O₇S
  • MDL Number:MFCD00005868
  • Molecular Weight:386.38 g/mol
  • Storage Temperature:Ambient
  • EINECS:204-088-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications