Orders LinkContactUsLinkComponent
2,4-Dimethylthiophenol 95%
2,4-Dimethylthiophenol 95%
Glass bottle for solids, 5 g
  B23161.06
2,4-Dimethylthiophenol 95%
  B23161.06
 :  13616-82-5
2,4-Dimethylthiophenol 95%
2,4-Dimethylthiophenol 95%
Glass bottle for solids, 5 g
 :  13616-82-5
94,70 €
1 * 5 g
Purity: 95% CAS No.: 13616-82-5 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 87°(189°F) Refractive Index: 1.5690 Odour: Unpleasant Boiling Point: 207-208° UN No.: 2810 MDL No.: MFCD00010019
 :  2,4-Dimethylbenzene-1-thiol, 1,3-Dimethyl-4-mercaptobenzene, 4-Mercapto-m-xylene, 2,4-Dimethylbenzenethiol
  •  :13616-82-5
  •  :C₈H₁₀S
  •  :MFCD00010019
  •  :138.23 g/mol
  •  :207…208 °C
  •  :1.020
  •  :87 °C (189 °F)
  •  :Ambient
  •  :237-100-0
  •  :2810
  •  :6.1,III
  •  :Density: 1.027 g/cm³ (20 °C)
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g