Isobutanol ≥99%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Isobutanol ≥99%
Catalog # B23091.AP
Supplier: Thermo Fisher Scientific
CAS Number: 78-83-1
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 78-83-1 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 28°(82°F) Refractive Index: 1.3960 Odour: Sweet pH: 7 Melting Point: -108° Boiling Point: 107-108° Beilstein Reference: 1730878 Merck Reference: 14,5131 UN No.: 1212 MDL No.: MFCD00004740
Synonyms: iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
- CAS Number:78-83-1
- Formula:(CH₃)₂CHCH₂OH
- MDL Number:MFCD00004740
- Molecular Weight:74.12 g/mol
- Merck Index:14,5131
- Boiling Pt:107…108 °C
- Melting Pt:–108 °C
- Density:0.802
- Flash Pt:28 °C (82 °F)
- Storage Temperature:Ambient
- EINECS:201-148-0
- UN:1212
- ADR:3,III
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Cat. No.:B23091.AP
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



