Order Entry
Export
Orders LinkContactUsLinkComponent
Isobutanol ≥99%

Isobutanol ≥99%

Glass bottle, 2.500 mL
Catalog # B23091.0F
Corrosive
Flammable
Irritant
Isobutanol ≥99%
Catalog # B23091.0F
CAS Number:  78-83-1
Isobutanol ≥99%
Isobutanol ≥99%

Glass bottle, 2.500 mL

CAS Number:  78-83-1
Price
118,00 €
Log in for availability
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 78-83-1 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 28°(82°F) Refractive Index: 1.3960 Odour: Sweet pH: 7 Melting Point: -108° Boiling Point: 107-108° Beilstein Reference: 1730878 Merck Reference: 14,5131 UN No.: 1212 MDL No.: MFCD00004740

Synonyms:  iso-Butyl alcohol, Isobutylalcohol, iso-Butanol, 2-Methyl-1-propanol
  • CAS Number:78-83-1
  • Formula:(CH₃)₂CHCH₂OH
  • MDL Number:MFCD00004740
  • Molecular Weight:74.12 g/mol
  • Merck Index:14,5131
  • Boiling Pt:107…108 °C
  • Melting Pt:–108 °C
  • Density:0.802
  • Flash Pt:28 °C (82 °F)
  • Storage Temperature:Ambient
  • EINECS:201-148-0
  • UN:1212
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2.500 mL

Specifications