3-(Dimethylamino)phenol ≥97%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
3-(Dimethylamino)phenol ≥97%
Catalog # B23067.22
Supplier: Thermo Fisher Scientific
CAS Number: 99-07-0
Jump to:
Product Details & Documents
Purity: 97+% CAS No.: 99-07-0 Molecular Formula: C8H11NO Formula Weight: 137.18 Flash Point: 148°(298°F) Odour: Pungent Boiling Point: 267° Beilstein Reference: 774631 MDL No.: MFCD00002264
Synonyms: N,N-Dimethyl-3-hydroxyaniline, 3-Dimethylaminophenol
- CAS Number:99-07-0
- Formula:C₈H₁₁NO
- MDL Number:MFCD00002264
- Molecular Weight:137.18 g/mol
- Boiling Pt:267 °C
- Melting Pt:80…84 °C
- Flash Pt:148 °C (298 °F)
- Storage Temperature:Ambient
- EINECS:202-727-0
Documents
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



