Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,5-Diaminopentane 98%

Glass bottle for solids, 25 g
Catalog # B23039.14
Corrosive
1,5-Diaminopentane 98%
Catalog # B23039.14
CAS Number:  462-94-2
1,5-Diaminopentane 98%
1,5-Diaminopentane 98%

Glass bottle for solids, 25 g

CAS Number:  462-94-2
Price
475,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 462-94-2 Molecular Formula: C5H14N2 Formula Weight: 102.18 Refractive Index: 1.4585 Odour: Amine-like Boiling Point: 178-180° Merck Reference: 14,1609 UN No.: 2735 MDL No.: MFCD00008239

Synonyms:  1,5-Pentanediamine, Pentamethylendiamine, Cadaverine, Cadavarine
  • CAS Number:462-94-2
  • Formula:C₅H₁₄N₂
  • MDL Number:MFCD00008239
  • Molecular Weight:102.18 g/mol
  • Merck Index:14,1609
  • Boiling Pt:178…180 °C
  • Melting Pt:9 °C
  • Density:0.873
  • Storage Temperature:Ambient
  • EINECS:207-329-0
  • ADR:8,II
  •  :Density: 0.9174 g/cm³ (0 °C)
  • UN:2735
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications