Order Entry
Export
Orders LinkContactUsLinkComponent
3,3-Diphenylpropylamine 97%

3,3-Diphenylpropylamine 97%

Glass bottle for solids, 25 g
Catalog # B22953.14
Irritant
3,3-Diphenylpropylamine 97%
Catalog # B22953.14
CAS Number:  5586-73-2
3,3-Diphenylpropylamine 97%
3,3-Diphenylpropylamine 97%

Glass bottle for solids, 25 g

CAS Number:  5586-73-2
Price
95,60 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 5586-73-2 Molecular Formula: (C6H5)2CHCH2CH2NH2 Formula Weight: 211.31 Flash Point: 110°(230°F) Refractive Index: 1.5830 Melting Point: 24-29° Boiling Point: 166°/2mm Beilstein Reference: 1958595 MDL No.: MFCD00008202

Synonyms:  3,3-Diphenylpropan-1-amine
  • CAS Number:5586-73-2
  • Formula:C₁₅H₁₇N
  • MDL Number:MFCD00008202
  • Molecular Weight:211.31 g/mol
  • Boiling Pt:166 °C (2 mmHg)
  • Melting Pt:24…29 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:226-984-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications