2,3-Dichlorophenol ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98+% CAS No.: 576-24-9 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Flash Point: 115°(239°F) Odour: Phenol-like Boiling Point: 206° Beilstein Reference: 2043615 UN No.: 2020 MDL No.: MFCD00002160
- CAS Number:576-24-9
- Formula:C₆H₄Cl₂O
- MDL Number:MFCD00002160
- Molecular Weight:163 g/mol
- Boiling Pt:206 °C
- Melting Pt:57…60 °C
- Density:1.4 g/cm³ (25 °C)
- Flash Pt:115 °C (239 °F)
- Storage Temperature:Ambient
- EINECS:209-399-8
- ADR:6.1,III
- :Density: 1.4 g/cm³ (25 °C)
- UN:2020
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



