Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dichlorophenol ≥98%

2,3-Dichlorophenol ≥98%

Glass bottle for solids, 100 g
Catalog # B22943.22
Irritant
2,3-Dichlorophenol ≥98%
Catalog # B22943.22
CAS Number:  576-24-9
2,3-Dichlorophenol ≥98%
2,3-Dichlorophenol ≥98%

Glass bottle for solids, 100 g

CAS Number:  576-24-9
Price
77,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 98+% CAS No.: 576-24-9 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Flash Point: 115°(239°F) Odour: Phenol-like Boiling Point: 206° Beilstein Reference: 2043615 UN No.: 2020 MDL No.: MFCD00002160

  • CAS Number:576-24-9
  • Formula:C₆H₄Cl₂O
  • MDL Number:MFCD00002160
  • Molecular Weight:163 g/mol
  • Boiling Pt:206 °C
  • Melting Pt:57…60 °C
  • Density:1.4 g/cm³ (25 °C)
  • Flash Pt:115 °C (239 °F)
  • Storage Temperature:Ambient
  • EINECS:209-399-8
  • ADR:6.1,III
  •  :Density: 1.4 g/cm³ (25 °C)
  • UN:2020
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications