Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl pyrocarbonate ≥97%

Diethyl pyrocarbonate ≥97%

Glass bottle for solids, 25 g
Catalog # B22753.14
Irritant
Diethyl pyrocarbonate ≥97%
Catalog # B22753.14
CAS Number:  1609-47-8
Diethyl pyrocarbonate ≥97%
Diethyl pyrocarbonate ≥97%

Glass bottle for solids, 25 g

CAS Number:  1609-47-8
Price
104,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 97% CAS No.: 1609-47-8 Molecular Formula: C6H10O5 Formula Weight: 162.14 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Physical Form: Clear liquid Density: 1.101 Refractive Index: 1.3980 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 93-94°/18mm Beilstein Reference: 637031 Merck Reference: 14,7998 MDL No.: MFCD00009106

Ribonuclease inhibitor

Synonyms:  DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
  • CAS Number:1609-47-8
  • Formula:O(COOC₂H₅)₂
  • MDL Number:MFCD00009106
  • Molecular Weight:162.14 g/mol
  • Merck Index:14,7998
  • Boiling Pt:93…94 °C (18 mmHg)
  • Density:1.101
  • Flash Pt:69 °C (156 °F)
  • Storage Temperature:Refrigerator
  • EINECS:216-542-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications