Diethyl pyrocarbonate ≥97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Diethyl pyrocarbonate ≥97%
Catalog # B22753.06
Supplier: Thermo Fisher Scientific
CAS Number: 1609-47-8
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 1609-47-8 Molecular Formula: C6H10O5 Formula Weight: 162.14 Storage Temperature: 2°C to 8°C Flash Point: 69°(156°F) Physical Form: Clear liquid Density: 1.101 Refractive Index: 1.3980 Sensitivity: Moisture Sensitive Appearance: Colourless Boiling Point: 93-94°/18mm Beilstein Reference: 637031 Merck Reference: 14,7998 MDL No.: MFCD00009106
Ribonuclease inhibitor
Synonyms: DEPC (Diethylpyrocarbonate), Diethyl dicarbonate, DEPC
- CAS Number:1609-47-8
- Formula:O(COOC₂H₅)₂
- MDL Number:MFCD00009106
- Molecular Weight:162.14 g/mol
- Merck Index:14,7998
- Boiling Pt:93…94 °C (18 mmHg)
- Density:1.101
- Flash Pt:69 °C (156 °F)
- Storage Temperature:Refrigerator
- EINECS:216-542-8
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



