Orders LinkContactUsLinkComponent
3,5-Dichlorobenzhydrazide ≥97%
Glass bottle for solids, 1 g
  B22383.03
3,5-Dichlorobenzhydrazide ≥97%
  B22383.03
 :  62899-78-9
3,5-Dichlorobenzhydrazide ≥97%
3,5-Dichlorobenzhydrazide ≥97%
Glass bottle for solids, 1 g
 :  62899-78-9
34,80 €
1 * 1 g
Purity: 97% CAS No.: 62899-78-9 Molecular Formula: C7H6Cl2N2O Formula Weight: 205.04 Melting Point: 205-207° MDL No.: MFCD00833411
 :  3,5-Dichlorobenzoic hydrazide
  •  :62899-78-9
  •  :C₇H₆Cl₂N₂O
  •  :MFCD00833411
  •  :205.04 g/mol
  •  :205…207 °C
  •  :Ambient
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g