Order Entry
Export
Orders LinkContactUsLinkComponent
2,3-Dimethylbutane ≥99%

2,3-Dimethylbutane ≥99%

Glass bottle for solids, 5 g
Catalog # B22223.06
Flammable
Health
Irritant
Environment
2,3-Dimethylbutane ≥99%
Catalog # B22223.06
CAS Number:  79-29-8
2,3-Dimethylbutane ≥99%
2,3-Dimethylbutane ≥99%

Glass bottle for solids, 5 g

CAS Number:  79-29-8
Price
38,20 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 99% CAS No.: 79-29-8 Molecular Formula: C6H14 Formula Weight: 86.18 Flash Point: -29°(-20°F) Refractive Index: 1.375 Odour: Mild Melting Point: -129° Boiling Point: 57-59° Beilstein Reference: 1730737 UN No.: 2457 MDL No.: MFCD00008925

  • CAS Number:79-29-8
  • Formula:C₆H₁₄
  • MDL Number:MFCD00008925
  • Molecular Weight:86.18 g/mol
  • Boiling Pt:57…59 °C
  • Melting Pt:–129 °C
  • Density:0.662
  • Flash Pt:–29 °C (–20 °F)
  • Storage Temperature:Ambient
  • EINECS:201-193-6
  • UN:2457
  • ADR:3,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications