Order Entry
Export
Orders LinkContactUsLinkComponent
2'-Hydroxy-4'-methoxyacetophenone ≥99%

2'-Hydroxy-4'-methoxyacetophenone ≥99%

Glass bottle for solids, 100 g
Catalog # B22173.22
Irritant
2'-Hydroxy-4'-methoxyacetophenone ≥99%
Catalog # B22173.22
CAS Number:  552-41-0
2'-Hydroxy-4'-methoxyacetophenone ≥99%
2'-Hydroxy-4'-methoxyacetophenone ≥99%

Glass bottle for solids, 100 g

CAS Number:  552-41-0
Price
862,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 552-41-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 1282794 MDL No.: MFCD00008730

Synonyms:  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  • CAS Number:552-41-0
  • Formula:C₉H₁₀O₃
  • MDL Number:MFCD00008730
  • Molecular Weight:166.18 g/mol
  • Boiling Pt:154 °C (20 mmHg)
  • Melting Pt:48…50 °C
  • Density:1.1604 g/cm³ (40 °C)
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:209-012-2
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications