Orders LinkContactUsLinkComponent
2'-Hydroxy-4'-methoxyacetophenone ≥99%
Glass bottle for solids, 5 g
  B22173.06
2'-Hydroxy-4'-methoxyacetophenone ≥99%
  B22173.06
 :  552-41-0
2'-Hydroxy-4'-methoxyacetophenone ≥99%
2'-Hydroxy-4'-methoxyacetophenone ≥99%
Glass bottle for solids, 5 g
 :  552-41-0
54,00 €
1 * 5 g
Purity: 99% CAS No.: 552-41-0 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 48-50° Beilstein Reference: 1282794 MDL No.: MFCD00008730
 :  1-(2-Hydroxy-4-methoxyphenyl)ethanone, Paeonol
  •  :552-41-0
  •  :C₉H₁₀O₃
  •  :MFCD00008730
  •  :166.18 g/mol
  •  :154 °C (20 mmHg)
  •  :48…50 °C
  •  :1.1604 g/cm³ (40 °C)
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :209-012-2
  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g