Orders LinkContactUsLinkComponent
1,6-Diiodoperfluorohexane ≥97%
Glass bottle for solids, 25 g
  B21654.14
1,6-Diiodoperfluorohexane ≥97%
  B21654.14
 :  375-80-4
1,6-Diiodoperfluorohexane ≥97%
1,6-Diiodoperfluorohexane ≥97%
Glass bottle for solids, 25 g
 :  375-80-4
489,00 €
1 * 25 g
Purity: 97% CAS No.: 375-80-4 Molecular Formula: I(CF2)6I Formula Weight: 553.86 Refractive Index: 1.3980 Melting Point: 25° Boiling Point: 115°/100mm Beilstein Reference: 1822186 MDL No.: MFCD00042264
 :  Perfluoro-1,6-diiodohexane
  •  :375-80-4
  •  :C₆F₁₂I₂
  •  :MFCD00042264
  •  :553.86 g/mol
  •  :115 °C (100 mmHg)
  •  :25 °C
  •  :2.357
  •  :Ambient
  •  :206-794-7
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g