Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,6-Diiodoperfluorohexane ≥97%

Glass bottle for solids, 5 g
Catalog # B21654.06
1,6-Diiodoperfluorohexane ≥97%
Catalog # B21654.06
CAS Number:  375-80-4
1,6-Diiodoperfluorohexane ≥97%
1,6-Diiodoperfluorohexane ≥97%

Glass bottle for solids, 5 g

CAS Number:  375-80-4
Price
112,00 €
Log in for availability
1 * 5 g
Product Details & Documents

Purity: 97% CAS No.: 375-80-4 Molecular Formula: I(CF2)6I Formula Weight: 553.86 Refractive Index: 1.3980 Melting Point: 25° Boiling Point: 115°/100mm Beilstein Reference: 1822186 MDL No.: MFCD00042264

Synonyms:  Perfluoro-1,6-diiodohexane
  • CAS Number:375-80-4
  • Formula:C₆F₁₂I₂
  • MDL Number:MFCD00042264
  • Molecular Weight:553.86 g/mol
  • Boiling Pt:115 °C (100 mmHg)
  • Melting Pt:25 °C
  • Density:2.357
  • Storage Temperature:Ambient
  • EINECS:206-794-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications