Order Entry
Export
Orders LinkContactUsLinkComponent
10-Methylphenothiazine ≥98%

10-Methylphenothiazine ≥98%

Glass bottle for solids, 25 g
Catalog # B21648.14
Irritant
10-Methylphenothiazine ≥98%
Catalog # B21648.14
CAS Number:  1207-72-3
10-Methylphenothiazine ≥98%
10-Methylphenothiazine ≥98%

Glass bottle for solids, 25 g

CAS Number:  1207-72-3
Price
227,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 1207-72-3 Molecular Formula: C13H11NS Formula Weight: 213.30 Melting Point: 100-103° MDL No.: MFCD00041836

  • CAS Number:1207-72-3
  • Formula:C₁₃H₁₁NS
  • MDL Number:MFCD00041836
  • Molecular Weight:213.3 g/mol
  • Boiling Pt:342 °C (1013 hPa)
  • Melting Pt:100…103 °C
  • Storage Temperature:Ambient
  • EINECS:214-896-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications