Order Entry
Export
Orders LinkContactUsLinkComponent
Calmagite

Calmagite

Glass bottle for solids, 25 g
Catalog # B21482.14
Irritant
Calmagite
Catalog # B21482.14
CAS Number:  3147-14-6
Calmagite
Calmagite

Glass bottle for solids, 25 g

CAS Number:  3147-14-6
Price
147,00 €
Log in for availability
1 * 25 g
Product Details & Documents

CAS No.: 3147-14-6 Molecular Formula: C17H14N2O5S Formula Weight: 358.37 Melting Point: 330° Beilstein Reference: 708596 Merck Reference: 14,1718 MDL No.: MFCD00011656

  • CAS Number:3147-14-6
  • Formula:C₁₇H₁₄N₂O₅S
  • MDL Number:MFCD00011656
  • Molecular Weight:358.37 g/mol
  • Merck Index:14,1718
  • Melting Pt:>330 °C
  • Storage Temperature:Ambient
  • EINECS:221-563-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications