Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Dihydroxynaphthalene ≥98%

Glass bottle for solids, 25 g
Catalog # B21465.14
Irritant
2,6-Dihydroxynaphthalene ≥98%
Catalog # B21465.14
CAS Number:  581-43-1
2,6-Dihydroxynaphthalene ≥98%
2,6-Dihydroxynaphthalene ≥98%

Glass bottle for solids, 25 g

CAS Number:  581-43-1
Price
323,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 581-43-1 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 223-225° Beilstein Reference: 1238082

Synonyms:  Naphthalene-2,6-diol
  • CAS Number:581-43-1
  • Formula:C₁₀H₈O₂
  • MDL Number:MFCD00004082
  • Molecular Weight:160.17 g/mol
  • Boiling Pt:375 °C (1013 hPa)
  • Melting Pt:223…225 °C
  • Density:1.33 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-465-6
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications