3-Bromophenetole ≥98%
Glass bottle for solids, 50 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 2655-84-7 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 83°(182°F) Refractive Index: 1.5480 Boiling Point: 231° MDL No.: MFCD00274287
Synonyms: 3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
- CAS Number:2655-84-7
- Formula:C₈H₉BrO
- MDL Number:MFCD00274287
- Molecular Weight:201.06 g/mol
- Boiling Pt:231 °C
- Density:1.41
- Flash Pt:83 °C (182 °F)
- Storage Temperature:Ambient
- EINECS:220-186-9
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:50 g



