Order Entry
Export
Orders LinkContactUsLinkComponent
3-Bromophenetole ≥98%

3-Bromophenetole ≥98%

Glass bottle for solids, 10 g
Catalog # B21418.09
3-Bromophenetole ≥98%
Catalog # B21418.09
CAS Number:  2655-84-7
3-Bromophenetole ≥98%
3-Bromophenetole ≥98%

Glass bottle for solids, 10 g

CAS Number:  2655-84-7
Price
60,50 €
Log in for availability
1 * 10 g
Product Details & Documents

Purity: 98% CAS No.: 2655-84-7 Molecular Formula: C8H9BrO Formula Weight: 201.06 Flash Point: 83°(182°F) Refractive Index: 1.5480 Boiling Point: 231° MDL No.: MFCD00274287

Synonyms:  3-Bromophenyl ethyl ether, 1-Bromo-3-ethoxybenzene
  • CAS Number:2655-84-7
  • Formula:C₈H₉BrO
  • MDL Number:MFCD00274287
  • Molecular Weight:201.06 g/mol
  • Boiling Pt:231 °C
  • Density:1.41
  • Flash Pt:83 °C (182 °F)
  • Storage Temperature:Ambient
  • EINECS:220-186-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications