Order Entry
Export
Orders LinkContactUsLinkComponent
 

3,4-Dimethylbenzophenone ≥99%

Glass bottle for solids, 100 g
Catalog # B20965.22
3,4-Dimethylbenzophenone ≥99%
Catalog # B20965.22
CAS Number:  2571-39-3
3,4-Dimethylbenzophenone ≥99%
3,4-Dimethylbenzophenone ≥99%

Glass bottle for solids, 100 g

CAS Number:  2571-39-3
Price
149,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99% CAS No.: 2571-39-3 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 335° Beilstein Reference: 1952043 MDL No.: MFCD00008525

Synonyms:  (3,4-Dimethylphenyl)(phenyl)methanone
  • CAS Number:2571-39-3
  • Formula:C₁₅H₁₄O
  • MDL Number:MFCD00008525
  • Molecular Weight:210.28 g/mol
  • Boiling Pt:335 °C
  • Melting Pt:44…47 °C
  • Flash Pt:>110 °C (230 °F)
  • Storage Temperature:Ambient
  • EINECS:219-916-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications