Orders LinkContactUsLinkComponent
3,4-Dimethylbenzophenone ≥99%
Glass bottle for solids, 25 g
  B20965.14
3,4-Dimethylbenzophenone ≥99%
  B20965.14
 :  2571-39-3
3,4-Dimethylbenzophenone ≥99%
3,4-Dimethylbenzophenone ≥99%
Glass bottle for solids, 25 g
 :  2571-39-3
45,90 €
1 * 25 g
Purity: 99% CAS No.: 2571-39-3 Molecular Formula: C15H14O Formula Weight: 210.28 Flash Point: 110°(230°F) Melting Point: 44-47° Boiling Point: 335° Beilstein Reference: 1952043 MDL No.: MFCD00008525
 :  (3,4-Dimethylphenyl)(phenyl)methanone
  •  :2571-39-3
  •  :C₁₅H₁₄O
  •  :MFCD00008525
  •  :210.28 g/mol
  •  :335 °C
  •  :44…47 °C
  •  :>110 °C (230 °F)
  •  :Ambient
  •  :219-916-9
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g