Order Entry
Export
Orders LinkContactUsLinkComponent
3,5-Dichlorosalicylaldehyde ≥98%

3,5-Dichlorosalicylaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # B20941.14
Irritant
3,5-Dichlorosalicylaldehyde ≥98%
Catalog # B20941.14
CAS Number:  90-60-8
3,5-Dichlorosalicylaldehyde ≥98%
3,5-Dichlorosalicylaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  90-60-8
Price
129,00 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 90-60-8 Molecular Formula: C7H4Cl2O2 Formula Weight: 191.01 Melting Point: 94-97° Beilstein Reference: 973391

Synonyms:  3,5-Dichloro-2-hydroxybenzaldehyde
  • CAS Number:90-60-8
  • Formula:C₇H₄Cl₂O₂
  • MDL Number:MFCD00003320
  • Molecular Weight:191.01 g/mol
  • Boiling Pt:723 °C (1013 hPa)
  • Melting Pt:94…97 °C
  • Storage Temperature:Ambient
  • EINECS:202-005-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications