Order Entry
Export
Orders LinkContactUsLinkComponent
 

Palmitic acid 95%

Glass bottle for solids, 2.500 g
Catalog # B20322.0E
Palmitic acid 95%
Catalog # B20322.0E
CAS Number:  57-10-3
Palmitic acid 95%
Palmitic acid 95%

Glass bottle for solids, 2.500 g

CAS Number:  57-10-3
Price
112,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 95% CAS No.: 57-10-3 Molecular Formula: C16H32O2 Formula Weight: 256.43 Flash Point: 206°(402°F) Refractive Index: 1.4273 Odour: Odourless Melting Point: 62-65° Boiling Point: 215°/15mm Beilstein Reference: 607489 Merck Reference: 14,6996 MDL No.: MFCD00002747

Synonyms:  Hexadecanoic acid, Hexadecylic acid
  • CAS Number:57-10-3
  • Formula:C₁₆H₃₂O₂
  • MDL Number:MFCD00002747
  • Molecular Weight:256.43 g/mol
  • Merck Index:14,6996
  • Boiling Pt:215 °C (15 mmHg)
  • Melting Pt:62…65 °C
  • Density:0.853
  • Flash Pt:206 °C (402 °F)
  • Storage Temperature:Ambient
  • EINECS:200-312-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications