2,4-Dihydroxybenzaldehyde ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
2,4-Dihydroxybenzaldehyde ≥98%
Catalog # A12868.36
Supplier: Thermo Fisher Scientific
CAS Number: 95-01-2
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 95-01-2 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 134-138° Boiling Point: 220°/22mm Merck Reference: 14,8156 MDL No.: MFCD00011686
- CAS Number:95-01-2
- Formula:C₇H₆O₃
- MDL Number:MFCD00011686
- Molecular Weight:138.12 g/mol
- Merck Index:14,8156
- Boiling Pt:220 °C (22 mmHg)
- Melting Pt:134…138 °C
- Density:1.448 g/cm³ (–93 °C)
- Storage Temperature:Ambient
- EINECS:202-383-1
Documents
Certificates
Search by Lot NumberALFAA12868.14_TDS_EN
Product specification
Organic Building Blocks and Intermediates
Application Notes
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



