Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,4-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g
Catalog # A12868.14
Irritant
2,4-Dihydroxybenzaldehyde ≥98%
Catalog # A12868.14
CAS Number:  95-01-2
2,4-Dihydroxybenzaldehyde ≥98%
2,4-Dihydroxybenzaldehyde ≥98%

Glass bottle for solids, 25 g

CAS Number:  95-01-2
Price
61,20 €
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 95-01-2 Molecular Formula: C7H6O3 Formula Weight: 138.12 Melting Point: 134-138° Boiling Point: 220°/22mm Merck Reference: 14,8156 MDL No.: MFCD00011686

  • CAS Number:95-01-2
  • Formula:C₇H₆O₃
  • MDL Number:MFCD00011686
  • Molecular Weight:138.12 g/mol
  • Merck Index:14,8156
  • Boiling Pt:220 °C (22 mmHg)
  • Melting Pt:134…138 °C
  • Density:1.448 g/cm³ (–93 °C)
  • Storage Temperature:Ambient
  • EINECS:202-383-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications