Order Entry
Export
Orders LinkContactUsLinkComponent
Anthranilic acid ≥98%

Anthranilic acid ≥98%

Glass bottle for solids, 1.000 g
Catalog # A12715.0B
Irritant
Anthranilic acid ≥98%
Catalog # A12715.0B
CAS Number:  118-92-3
Anthranilic acid ≥98%
Anthranilic acid ≥98%

Glass bottle for solids, 1.000 g

CAS Number:  118-92-3
Price
108,00 €
Log in for availability
1 * 1.000 g
Product Details & Documents
Synonyms:  2-Aminobenzoic acid
  • CAS Number:118-92-3
  • Formula:C₇H₇NO₂
  • MDL Number:MFCD00007712
  • Molecular Weight:137.14 g/mol
  • Merck Index:14,422
  • Boiling Pt:312 °C (1013 hPa)
  • Melting Pt:145…148 °C
  • Density:1.412
  • Flash Pt:150 °C (302 °F)
  •  :Density: 1.362 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:204-287-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1.000 g

Specifications