Calcium hydroxide 95%
Glass bottle for solids, 250 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Slaked lime, Calcium dihydroxide
- CAS Number:1305-62-0
- Formula:Ca(OH)₂
- MDL Number:MFCD00010901
- Molecular Weight:74.09 g/mol
- Merck Index:14,1673
- Melting Pt:550 °C
- Density:2.24
- Storage Temperature:Ambient
- EINECS:215-137-3
- UN:3262
- ADR:8,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g



