Order Entry
Export
Orders LinkContactUsLinkComponent
 

Calcium hydroxide 95%

Glass bottle for solids, 5.000 g
Catalog # A12650.0I
Corrosive
Irritant
Calcium hydroxide 95%
Catalog # A12650.0I
CAS Number:  1305-62-0
Calcium hydroxide 95%
Calcium hydroxide 95%

Glass bottle for solids, 5.000 g

CAS Number:  1305-62-0
Price
151,00 €
Log in for availability
1 * 5.000 g
Product Details & Documents
Synonyms:  Slaked lime, Calcium dihydroxide
  • CAS Number:1305-62-0
  • Formula:Ca(OH)₂
  • MDL Number:MFCD00010901
  • Molecular Weight:74.09 g/mol
  • Merck Index:14,1673
  • Melting Pt:550 °C
  • Density:2.24
  • Storage Temperature:Ambient
  • EINECS:215-137-3
  • UN:3262
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5.000 g

Specifications