Triethylamine 99%
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 121-44-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -11°(12°F) Refractive Index: 1.4005 Odour: Ammonia-like Melting Point: -115° Boiling Point: 89-90° Beilstein Reference: 1843166 Merck Reference: 14,9666 UN No.: 1296 MDL No.: MFCD00009051
Synonyms: (Diethylamino)ethane, TEA
- CAS Number:121-44-8
- Formula:(C₂H₅)₃N
- MDL Number:MFCD00009051
- Molecular Weight:101.19 g/mol
- Merck Index:14,9666
- Boiling Pt:89…90 °C
- Melting Pt:–115 °C
- Density:0.726
- Flash Pt:–11 °C (–12 °F)
- Storage Temperature:Ambient
- EINECS:204-469-4
- UN:1296
- ADR:3,II
Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:250 mL



