Benzil ≥98%
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 134-81-6 Molecular Formula: C14H10O2 Formula Weight: 210.23 Melting Point: 94-96° Boiling Point: 346-348° Merck Reference: 14,1078 MDL No.: MFCD00003080
Synonyms: 1,2-Diphenyl-1,2-ethanedione
- CAS Number:134-81-6
- Formula:(C₆H₅)C(O)C(O)(C₆H₅)
- MDL Number:MFCD00003080
- Molecular Weight:210.23 g/mol
- Merck Index:14,1078
- Boiling Pt:346…348 °C
- Melting Pt:94…96 °C
- Density:1.521
- Storage Temperature:Ambient
- EINECS:205-157-0
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



