Order Entry
Export
Orders LinkContactUsLinkComponent
 

Benzil ≥98%

Glass bottle for solids, 2.500 g
Catalog # A12579.0E
Irritant
Benzil ≥98%
Catalog # A12579.0E
CAS Number:  134-81-6
Benzil ≥98%
Benzil ≥98%

Glass bottle for solids, 2.500 g

CAS Number:  134-81-6
Price
301,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 98% CAS No.: 134-81-6 Molecular Formula: C14H10O2 Formula Weight: 210.23 Melting Point: 94-96° Boiling Point: 346-348° Merck Reference: 14,1078 MDL No.: MFCD00003080

Synonyms:  1,2-Diphenyl-1,2-ethanedione
  • CAS Number:134-81-6
  • Formula:(C₆H₅)C(O)C(O)(C₆H₅)
  • MDL Number:MFCD00003080
  • Molecular Weight:210.23 g/mol
  • Merck Index:14,1078
  • Boiling Pt:346…348 °C
  • Melting Pt:94…96 °C
  • Density:1.521
  • Storage Temperature:Ambient
  • EINECS:205-157-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications