Order Entry
Export
Orders LinkContactUsLinkComponent
 

Piperidine ≥99%

Glass bottle, 500 mL
Catalog # A12442.AP
Flammable
Toxic
Corrosive
Piperidine ≥99%
Catalog # A12442.AP
CAS Number:  110-89-4
Piperidine ≥99%
Piperidine ≥99%

Glass bottle, 500 mL

CAS Number:  110-89-4
Price
63,40 €
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 110-89-4 Molecular Formula: C5H11N Formula Weight: 85.15 Flash Point: 3°(37°F) Physical Form: Liquid Density: 0.862 Refractive Index: 1.4520 Sensitivity: Air Sensitive Appearance: Colourless Melting Point: -11° Boiling Point: 105-106° Beilstein Reference: 102438 Merck Reference: 14,7468 UN No.: 2401 MDL No.: MFCD00005979

Synonyms:  Hexahydropyridine, Hexazane, Pentamethyleneimine
  • CAS Number:110-89-4
  • Formula:C₅H₁₁N
  • MDL Number:MFCD00005979
  • Molecular Weight:85.15 g/mol
  • Merck Index:14,7468
  • Boiling Pt:105…106 °C
  • Melting Pt:–11 °C
  • Density:0.862
  • Flash Pt:3 °C (37 °F)
  • Storage Temperature:Ambient
  • EINECS:203-813-0
  • UN:2401
  • ADR:8,I
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications