Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,6-Hexanediol ≥97%

Glass bottle for solids, 2.500 g
Catalog # A12439.0E
1,6-Hexanediol ≥97%
Catalog # A12439.0E
CAS Number:  629-11-8
1,6-Hexanediol ≥97%
1,6-Hexanediol ≥97%

Glass bottle for solids, 2.500 g

CAS Number:  629-11-8
Price
177,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 97% CAS No.: 629-11-8 Molecular Formula: HO(CH2)6OH Formula Weight: 118.18 Flash Point: 147°(296°F) Physical Form: Cryst solid Density: 1.120 Sensitivity: Hygroscopic Appearance: White Melting Point: 40-43° Boiling Point: 248-250° Beilstein Reference: 1633461 Merck Reference: 14,4690 MDL No.: MFCD00002985

Synonyms:  Hexamethylene glycol, 1,6-Dihydroxy hexane
  • CAS Number:629-11-8
  • Formula:C₆H₁₄O₂
  • MDL Number:MFCD00002985
  • Molecular Weight:118.18 g/mol
  • Merck Index:14,4690
  • Boiling Pt:248…250 °C
  • Melting Pt:40…43 °C
  • Density:1.120
  • Flash Pt:147 °C (296 °F)
  • Storage Temperature:Ambient
  • EINECS:211-074-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications