Orders LinkContactUsLinkComponent
2,7-Dibromofluorene ≥98%
Glass bottle for solids, 100 g
  A12437.22
2,7-Dibromofluorene ≥98%
  A12437.22
 :  16433-88-8
2,7-Dibromofluorene ≥98%
2,7-Dibromofluorene ≥98%
Glass bottle for solids, 100 g
 :  16433-88-8
292,00 €
1 * 100 g
Purity: 98% CAS No.: 16433-88-8 Molecular Formula: C13H8Br2 Formula Weight: 324.03 Melting Point: 164-167° Beilstein Reference: 2049205
 :  2,7-Dibromo-9H-fluorene
  •  :16433-88-8
  •  :C₁₃H₈Br₂
  •  :MFCD00019048
  •  :324.01 g/mol
  •  :164…167 °C
  •  :Ambient
  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g